siRNA Details
| Gene Symbol: Zfp347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133390.1 | NP_596881.1 | 5 | 4671 | ||
| GenBank mRNA | AB047637.1 | - | 5 | 4671 | ||
| AB047638.1 | - | 3057 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp347| Gene Symbol: Zfp347 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133390.1 | NP_596881.1 | 5 | 4671 | ||
| GenBank mRNA | AB047637.1 | - | 5 | 4671 | ||
| AB047638.1 | - | 3057 | ||||