siRNA Details
| Gene Symbol: A1cf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133400.1 | NP_596891.1 | 5, 6 | 714 | ||
| XM_006231272.2 | XP_006231334.1 | 5, 6 | 885 | |||
| GenBank mRNA | AF290984.1 | - | 5, 6 | 714 | ||
| AF442133.1 | - | 5, 6 | 714 | |||
| AF442134.1 | - | 5, 6 | 714 | |||
| AY028945.1 | - | 4, 5 | 612 | |||

A1cf
Genomic Map