siRNA Details
| Gene Symbol: Necab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133415.1 | NP_596906.1 | 13 | 1220 | ||
| XM_006255703.2 | XP_006255765.1 | 11 | 2286 | |||
| GenBank mRNA | AF193757.1 | - | 13 | 1220 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Necab2| Gene Symbol: Necab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133415.1 | NP_596906.1 | 13 | 1220 | ||
| XM_006255703.2 | XP_006255765.1 | 11 | 2286 | |||
| GenBank mRNA | AF193757.1 | - | 13 | 1220 | ||