siRNA Details
| Gene Symbol: LOC103689943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006245686.2 | XP_006245748.1 | 3, 4 | 2070 | ||
| XM_006245687.2 | XP_006245749.1 | 3, 4 | 996 | |||
| XM_006245688.1 | XP_006245750.1 | 3, 4 | 848 | |||
| XM_008767447.1 | XP_008765669.1 | 3, 4 | 996 | |||
| XM_008767448.1 | XP_008765670.1 | 2, 3 | 1503 | |||
| Gene Symbol: Marf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133421.2 | NP_596912.1 | 3, 4 | 1022 | ||
| XM_003750779.3 | XP_003750827.1 | 3, 4 | 2069 | |||
| XM_006245854.2 | XP_006245916.1 | 3, 4 | 996 | |||
| XM_006245855.2 | XP_006245917.1 | 3, 4 | 848 | |||
| XM_008767442.1 | XP_008765664.1 | 2, 3 | 1503 | |||
| GenBank mRNA | AB012133.1 | - | 3, 4 | 1022 | ||

LOC103689943
Genomic Map