siRNA Details
| Gene Symbol: Akap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133515.2 | NP_598199.2 | 2 | 681 | ||
| GenBank mRNA | U67136.1 | - | 2 | 680 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Akap5| Gene Symbol: Akap5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133515.2 | NP_598199.2 | 2 | 681 | ||
| GenBank mRNA | U67136.1 | - | 2 | 680 | ||