siRNA Details
| Gene Symbol: Cd79b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133533.2 | NP_598217.1 | 2 | 148 | ||
| GenBank mRNA | BC088115.1 | - | 2 | 148 | 7325406 | |
| FQ226266.1 | - | 2 | 145 | |||
| FQ227061.1 | - | 2 | 143 | |||
| FQ227788.1 | - | 2 | 137 | |||
| FQ230261.1 | - | 2 | 139 | |||
| FQ232054.1 | - | 2 | 139 | |||
| FQ232680.1 | - | 2 | 139 | |||
| FQ233507.1 | - | 2 | 137 | |||
| FQ234082.1 | - | 2 | 122 | |||
| FQ234487.1 | - | 2 | 137 | |||

Cd79b
Genomic Map