siRNA Details
Gene Symbol: Ptpn21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133545.1 | NP_598229.1 | 2 | 349 | ||
GenBank mRNA | U17971.1 | - | 2 | 349 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Ptpn21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133545.1 | NP_598229.1 | 2 | 349 | ||
GenBank mRNA | U17971.1 | - | 2 | 349 |