siRNA Details
Gene Symbol: Dusp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133578.1 | NP_598262.1 | 4 | 1729 | ||
GenBank mRNA | AF013144.1 | - | 4 | 1729 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dusp5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133578.1 | NP_598262.1 | 4 | 1729 | ||
GenBank mRNA | AF013144.1 | - | 4 | 1729 |