siRNA Details
| Gene Symbol: Ppp6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133589.2 | NP_598273.2 | 6 | 646 | ||
| GenBank mRNA | BC078747.1 | - | 6 | 646 | 7131278 | |
| X77236.1 | - | 6 | 611 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ppp6c| Gene Symbol: Ppp6c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133589.2 | NP_598273.2 | 6 | 646 | ||
| GenBank mRNA | BC078747.1 | - | 6 | 646 | 7131278 | |
| X77236.1 | - | 6 | 611 | |||