siRNA Details
| Gene Symbol: Ptprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191945.1 | NP_001178874.1 | 11, 12 | 1931 | ||
| XM_008758887.1 | XP_008757109.1 | 14, 15 | 3484 | |||
| GenBank mRNA | D45413.1 | - | 3, 4 | 480 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprh| Gene Symbol: Ptprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191945.1 | NP_001178874.1 | 11, 12 | 1931 | ||
| XM_008758887.1 | XP_008757109.1 | 14, 15 | 3484 | |||
| GenBank mRNA | D45413.1 | - | 3, 4 | 480 | ||