siRNA Details
| Gene Symbol: Ptprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191945.1 | NP_001178874.1 | 12, 13 | 2027 | ||
| XM_008758887.1 | XP_008757109.1 | 15, 16 | 3580 | |||
| GenBank mRNA | D45413.1 | - | 4, 5 | 576 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ptprh| Gene Symbol: Ptprh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191945.1 | NP_001178874.1 | 12, 13 | 2027 | ||
| XM_008758887.1 | XP_008757109.1 | 15, 16 | 3580 | |||
| GenBank mRNA | D45413.1 | - | 4, 5 | 576 | ||