siRNA Details
| Gene Symbol: Kcnh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133610.2 | NP_598294.1 | 4 | 661 | ||
| GenBank mRNA | AF185637.1 | - | 4 | 620 | ||
| AJ250280.1 | - | 4 | 663 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Kcnh5| Gene Symbol: Kcnh5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133610.2 | NP_598294.1 | 4 | 661 | ||
| GenBank mRNA | AF185637.1 | - | 4 | 620 | ||
| AJ250280.1 | - | 4 | 663 | |||