siRNA Details
Gene Symbol: Hadhb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006239786.1 | XP_006239848.1 | 3 | 561 | ||
XM_006239788.1 | XP_006239850.1 | 3 | 217 | |||
XM_006239789.2 | XP_006239851.1 | 3 | 492 | |||
GenBank mRNA | BC060545.1 | - | 3 | 144 | 6918791 | |
D16479.1 | - | 2 | 108 | |||
FM050540.1 | - | 3 | 172 |
