siRNA Details
| Gene Symbol: PHB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144831.1 | NP_001138303.1 | 2, 3 | 430 | ||
| NM_001267700.1 | NP_001254629.1 | 2, 3 | 430 | |||
| GenBank mRNA | AF126021.1 | - | 2, 3 | 405 | ||
| AF150962.1 | - | 2, 3 | 343 | |||
| AF178980.1 | - | 2, 3 | 319 | |||
| AK298217.1 | - | 2, 3 | 350 | |||
| AK301316.1 | - | 2, 3 | 394 | |||
| BC013228.1 | - | 2, 3 | 313 | |||
| BC014766.1 | - | 2, 3 | 300 | 3452360 | ||
| BC110322.1 | - | 2, 3 | 430 | 5540527 | ||

PHB2
Genomic Map