siRNA Details
Gene Symbol: Glyatl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126278.1 | NP_001119750.1 | 2 | 117 | ||
GenBank mRNA | BC160862.1 | - | 2 | 117 | 7323546 | |
FQ210014.1 | - | 271 | ||||
FQ219712.1 | - | 2 | 120 |
Gene Symbol: Glyatl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134330.2 | NP_599157.2 | 3 | 156 | ||
GenBank mRNA | AB019693.1 | - | 222 | |||
BC078701.1 | - | 3 | 156 | 7130696 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
