siRNA Details
| Gene Symbol: Slc12a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134363.1 | NP_599190.1 | 3 | 333 | ||
| GenBank mRNA | EF641113.1 | - | 346 | |||
| U55816.1 | - | 3 | 333 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc12a5| Gene Symbol: Slc12a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134363.1 | NP_599190.1 | 3 | 333 | ||
| GenBank mRNA | EF641113.1 | - | 346 | |||
| U55816.1 | - | 3 | 333 | |||