siRNA Details
Gene Symbol: Trpm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134371.2 | NP_599198.2 | 26 | 4075 | ||
GenBank mRNA | AY072788.2 | - | 26 | 4075 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Trpm8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134371.2 | NP_599198.2 | 26 | 4075 | ||
GenBank mRNA | AY072788.2 | - | 26 | 4075 |