siRNA Details
| Gene Symbol: Ust5r | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134380.1 | NP_599207.1 | 8 | 1545 | ||
| XM_008760213.1 | XP_008758435.1 | 8 | 2158 | |||
| GenBank mRNA | AJ132858.1 | - | 8 | 1545 | ||
| BC081730.1 | - | 8 | 1864 | |||
| BC098905.1 | - | 8 | 1521 | 7325342 | ||

Ust5r
Genomic Map