siRNA Details
| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134401.2 | NP_599228.2 | 13 | 1756 | ||
| GenBank mRNA | BC089944.1 | - | 11 | 1287 | ||
| U78304.1 | - | 4 | 397 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Crtac1| Gene Symbol: Crtac1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134401.2 | NP_599228.2 | 13 | 1756 | ||
| GenBank mRNA | BC089944.1 | - | 11 | 1287 | ||
| U78304.1 | - | 4 | 397 | |||