siRNA Details
| Gene Symbol: SYNPO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001109974.2 | NP_001103444.1 | 2 | 619 | ||
| NM_001166208.1 | NP_001159680.1 | 3 | 1183 | |||
| NM_001166209.1 | NP_001159681.1 | 3 | 1116 | |||
| NM_007286.5 | NP_009217.3 | 2 | 619 | |||
| XM_005268370.1 | XP_005268427.1 | 2 | 562 | |||
| XM_005268371.1 | XP_005268428.1 | 2 | 482 | |||
| XM_006714755.3 | XP_006714818.1 | 3 | 1171 | |||
| XM_011537552.2 | XP_011535854.1 | 3 | 1168 | |||
| XM_017009007.1 | XP_016864496.1 | 3 | 1204 | |||
| XM_017009008.1 | XP_016864497.1 | 3 | 1143 | |||
| GenBank mRNA | AB028952.1 | - | 2 | 573 | ||
| AF499136.1 | - | 2 | 533 | |||
| AF499137.1 | - | 2 | 533 | |||
| AK127049.1 | - | 3 | 1116 | |||
| AK290178.1 | - | 2 | 602 | |||
| AL831818.1 | - | 3 | 1183 | |||
| BC142683.1 | - | 2 | 540 | 40148430 | ||
| BC146665.1 | - | 2 | 586 | 40148077 | ||
| BC152538.1 | - | 2 | 573 | |||
| Y11072.1 | - | 1 | 77 | |||

SYNPO
Genomic Map