siRNA Details
| Gene Symbol: Lphn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF063102.1 | - | 5 | 582 | ||
| AF081148.1 | - | 6 | 1028 | |||
| AF081149.1 | - | 6 | 1028 | |||
| AF081150.1 | - | 6 | 1028 | |||
| AF081151.1 | - | 6 | 1028 | |||
| AF081152.1 | - | 6 | 1028 | |||
| AF081153.1 | - | 6 | 1028 | |||
| CB579130.1 | - | 4 | 313 | |||

Lphn2
Genomic Map