siRNA Details
Gene Symbol: Rab3il1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134411.1 | NP_599238.1 | 2 | 284 | ||
GenBank mRNA | AY026049.1 | - | 2 | 284 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rab3il1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_134411.1 | NP_599238.1 | 2 | 284 | ||
GenBank mRNA | AY026049.1 | - | 2 | 284 |