siRNA Details
| Gene Symbol: Hspbap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134419.2 | NP_599246.2 | 2 | 137 | ||
| GenBank mRNA | AF168362.1 | - | 2 | 126 | ||
| BC091125.1 | - | 2 | 137 | 7366096 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hspbap1| Gene Symbol: Hspbap1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134419.2 | NP_599246.2 | 2 | 137 | ||
| GenBank mRNA | AF168362.1 | - | 2 | 126 | ||
| BC091125.1 | - | 2 | 137 | 7366096 | ||