siRNA Details
| Gene Symbol: Psmc3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134458.2 | NP_604453.2 | 5 | 478 | ||
| XM_006247249.2 | XP_006247311.1 | 5 | 1330 | |||
| GenBank mRNA | AF352812.1 | - | 5 | 478 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Psmc3ip| Gene Symbol: Psmc3ip | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134458.2 | NP_604453.2 | 5 | 478 | ||
| XM_006247249.2 | XP_006247311.1 | 5 | 1330 | |||
| GenBank mRNA | AF352812.1 | - | 5 | 478 | ||