siRNA Details
| Gene Symbol: Kif2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085369.1 | NP_001078838.1 | 3, 4 | 338 | ||
| NM_134472.3 | NP_608302.2 | 2, 3 | 223 | |||
| XM_006238632.2 | XP_006238694.1 | 3, 4 | 387 | |||
| XM_006238633.1 | XP_006238695.1 | 2, 3 | 151 | |||
| XM_006238634.2 | XP_006238696.1 | 2, 3 | 683 | |||
| XM_006238635.2 | XP_006238697.1 | 3, 4 | 560 | |||
| XM_006238636.2 | XP_006238698.1 | 3, 4 | 556 | |||
| GenBank mRNA | CO575506.1 | - | 2, 3 | 146 | ||

Kif2c
Genomic Map