siRNA Details
| Gene Symbol: Osgin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138504.2 | NP_612513.1 | 5, 6 | 750 | ||
| XM_006255706.2 | XP_006255768.1 | 8, 9 | 809 | |||
| XM_006255707.2 | XP_006255769.1 | 4, 5 | 845 | |||
| XM_008772594.1 | XP_008770816.1 | 3, 4 | 671 | |||
| GenBank mRNA | AF549441.1 | - | 5, 6 | 750 | ||
| AF549442.1 | - | 4, 5 | 651 | |||
| AY081218.1 | - | 3, 4 | 399 | |||
| BC091111.1 | - | 5, 6 | 746 | 7106386 | ||

Osgin1
Genomic Map