siRNA Details
| Gene Symbol: Osgin1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138504.2 | NP_612513.1 | 7 | 1008 | ||
| XM_006255706.2 | XP_006255768.1 | 10 | 1067 | |||
| XM_006255707.2 | XP_006255769.1 | 6 | 1103 | |||
| XM_008772594.1 | XP_008770816.1 | 5 | 929 | |||
| GenBank mRNA | AF549441.1 | - | 7 | 1008 | ||
| AF549442.1 | - | 6 | 909 | |||
| AY081218.1 | - | 5 | 657 | |||
| BC091111.1 | - | 7 | 1004 | 7106386 | ||

Osgin1
Genomic Map