siRNA Details
| Gene Symbol: Prpf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138523.2 | NP_612532.1 | 10 | 1449 | ||
| GenBank mRNA | AF244920.1 | - | 10 | 1445 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Prpf18| Gene Symbol: Prpf18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138523.2 | NP_612532.1 | 10 | 1449 | ||
| GenBank mRNA | AF244920.1 | - | 10 | 1445 | ||