siRNA Details
| Gene Symbol: Stambp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138531.2 | NP_612540.1 | 4, 5 | 504 | ||
| XM_006236726.2 | XP_006236788.1 | 5, 6 | 1171 | |||
| XM_006236727.2 | XP_006236789.1 | 6, 7 | 1289 | |||
| XM_008763045.1 | XP_008761267.1 | 4, 5 | 2832 | |||
| XM_008763046.1 | XP_008761268.1 | 5, 6 | 1852 | |||
| XM_008763047.1 | XP_008761269.1 | 5, 6 | 2269 | |||
| XM_008763048.1 | XP_008761270.1 | 4, 5 | 2151 | |||
| GenBank mRNA | AY083159.1 | - | 3, 4 | 328 | ||
| BC061711.1 | - | 4, 5 | 504 | 5597943 | ||

Stambp
Genomic Map