siRNA Details
| Gene Symbol: Fat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138544.1 | NP_612553.1 | 2 | 1314 | ||
| GenBank mRNA | AB076401.1 | - | 2 | 1314 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fat3| Gene Symbol: Fat3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138544.1 | NP_612553.1 | 2 | 1314 | ||
| GenBank mRNA | AB076401.1 | - | 2 | 1314 | ||