siRNA Details
| Gene Symbol: Vmp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138839.2 | NP_620194.1 | 12 | 1464 | ||
| XM_006247078.2 | XP_006247140.1 | 12 | 1942 | |||
| XM_006247079.2 | XP_006247141.1 | 13 | 1486 | |||
| GenBank mRNA | AF411216.1 | - | 12 | 1410 | ||
| BC061721.1 | - | 12 | 1464 | 5598561 | ||
| FQ220200.1 | - | 3 | 616 | |||
| FQ220288.1 | - | 3 | 630 | |||
| FQ220745.1 | - | 2 | 585 | |||
| FQ220965.1 | - | 3 | 616 | |||

Vmp1
Genomic Map