siRNA Details
| Gene Symbol: Prom2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138857.1 | NP_620212.1 | 5 | 779 | ||
| XM_006234947.2 | XP_006235009.1 | 5 | 1578 | |||
| GenBank mRNA | AF486828.1 | - | 5 | 779 | ||
| AF508942.1 | - | 5 | 761 | |||
| FQ229034.1 | - | 3 | 414 | |||

Prom2
Genomic Map