siRNA Details
| Gene Symbol: Slc9a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138858.1 | NP_620213.1 | 6 | 1099 | ||
| GenBank mRNA | AF100172.1 | - | 6 | 1099 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc9a5| Gene Symbol: Slc9a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138858.1 | NP_620213.1 | 6 | 1099 | ||
| GenBank mRNA | AF100172.1 | - | 6 | 1099 | ||