siRNA Details
| Gene Symbol: Ptgr1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138863.2 | NP_620218.1 | 5 | 291 | ||
| GenBank mRNA | BC089775.1 | - | 5 | 291 | 7307365 | |
| FQ218841.1 | - | 5 | 310 | |||
| FQ220898.1 | - | 5 | 312 | |||
| FQ221589.1 | - | 5 | 307 | |||
| FQ222466.1 | - | 5 | 309 | |||
| FQ229147.1 | - | 5 | 307 | |||
| FQ229679.1 | - | 5 | 307 | |||
| U66322.2 | - | 4 | 256 | |||

Ptgr1
Genomic Map