siRNA Details
| Gene Symbol: Cep19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138865.1 | NP_620220.1 | 4 | 580 | ||
| XM_008768696.1 | XP_008766918.1 | 4 | 759 | |||
| XM_008768697.1 | XP_008766919.1 | 4 | 745 | |||
| GenBank mRNA | AF146738.1 | - | 4 | 580 | ||
| FQ211926.1 | - | 4 | 597 | |||

Cep19
Genomic Map