siRNA Details
| Gene Symbol: Hyou1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001034028.1 | NP_001029200.1 | 4 | 294 | ||
| NM_138867.2 | NP_620222.2 | 4 | 446 | |||
| XM_006242884.2 | XP_006242946.1 | 4 | 391 | |||
| XM_006242885.2 | XP_006242947.1 | 4 | 327 | |||
| XM_008766143.1 | XP_008764365.1 | 4 | 373 | |||
| GenBank mRNA | BC065310.1 | - | 4 | 337 | 5621638 | |
| CB691248.1 | - | 4 | 302 | |||
| U41853.1 | - | 4 | 446 | |||

Hyou1
Genomic Map