siRNA Details
| Gene Symbol: Golgb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138885.1 | NP_620240.1 | 5, 6 | 695 | ||
| XM_006248379.2 | XP_006248441.1 | 5, 6 | 2565 | |||
| XM_006248380.2 | XP_006248442.1 | 4, 5 | 2413 | |||
| XM_008768698.1 | XP_008766920.1 | 4, 5 | 1758 | |||
| XM_008768699.1 | XP_008766921.1 | 4, 5 | 1758 | |||
| XM_008768700.1 | XP_008766922.1 | 4, 5 | 1758 | |||
| XM_008768701.1 | XP_008766923.1 | 4, 5 | 1758 | |||
| XM_008768702.1 | XP_008766924.1 | 3, 4 | 407 | |||
| XM_008768703.1 | XP_008766925.1 | 4, 5 | 1759 | |||
| GenBank mRNA | D25543.1 | - | 5, 6 | 695 | ||

Golgb1
Genomic Map