siRNA Details
| Gene Symbol: Gabrr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138897.1 | NP_620252.1 | 1 | 287 | ||
| GenBank mRNA | D50671.1 | - | 1 | 287 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gabrr3| Gene Symbol: Gabrr3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138897.1 | NP_620252.1 | 1 | 287 | ||
| GenBank mRNA | D50671.1 | - | 1 | 287 | ||