siRNA Details
| Gene Symbol: Pigl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138901.2 | NP_620256.1 | 3 | 416 | ||
| XM_008767767.1 | XP_008765989.1 | 3 | 412 | |||
| GenBank mRNA | BC074020.1 | - | 3 | 416 | 7104008 | |
| D88364.1 | - | 3 | 797 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pigl| Gene Symbol: Pigl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138901.2 | NP_620256.1 | 3 | 416 | ||
| XM_008767767.1 | XP_008765989.1 | 3 | 412 | |||
| GenBank mRNA | BC074020.1 | - | 3 | 416 | 7104008 | |
| D88364.1 | - | 3 | 797 | |||