siRNA Details
| Gene Symbol: Gls2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001270786.1 | NP_001257715.1 | 18 | 2283 | ||
| NM_001270787.1 | NP_001257716.1 | 18 | 2529 | |||
| NM_138904.2 | NP_620259.2 | 17 | 1978 | |||
| XM_006240728.2 | XP_006240790.1 | 17 | 2216 | |||
| XM_006240730.2 | XP_006240792.1 | 19 | 2108 | |||
| GenBank mRNA | BC089776.1 | - | 18 | 2122 | ||
| BC104712.1 | - | 18 | 2027 | |||
| FQ211735.1 | - | 18 | 2529 | |||
| J05499.1 | - | 17 | 1977 | |||

Gls2
Genomic Map