siRNA Details
Gene Symbol: Slc22a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138908.2 | NP_620263.1 | 3 | 610 | ||
GenBank mRNA | Y09945.1 | - | 3 | 612 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Slc22a25 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_138908.2 | NP_620263.1 | 3 | 610 | ||
GenBank mRNA | Y09945.1 | - | 3 | 612 |