siRNA Details
| Gene Symbol: Slc22a25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138908.2 | NP_620263.1 | 7 | 1266 | ||
| GenBank mRNA | Y09945.1 | - | 7 | 1268 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Slc22a25| Gene Symbol: Slc22a25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138908.2 | NP_620263.1 | 7 | 1266 | ||
| GenBank mRNA | Y09945.1 | - | 7 | 1268 | ||