siRNA Details
| Gene Symbol: Fnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138914.1 | NP_620269.1 | 992 | |||
| GenBank mRNA | AB073208.1 | - | 992 | |||
| AB126170.1 | - | 992 | ||||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fnbp1| Gene Symbol: Fnbp1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138914.1 | NP_620269.1 | 992 | |||
| GenBank mRNA | AB073208.1 | - | 992 | |||
| AB126170.1 | - | 992 | ||||