siRNA Details
| Gene Symbol: Zc3h14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001033951.1 | NP_001029123.1 | 2, 3 | 172 | ||
| NM_138920.1 | NP_620275.1 | 2, 3 | 172 | |||
| XM_008764738.1 | XP_008762960.1 | 2, 3 | 209 | |||
| XM_008764739.1 | XP_008762961.1 | 2, 3 | 209 | |||
| GenBank mRNA | AB032932.1 | - | 2, 3 | 172 | ||
| AB097075.1 | - | 2, 3 | 172 | |||
| AB097076.1 | - | 2, 3 | 172 | |||
| AB097077.1 | - | 2, 3 | 172 | |||
| BC087712.1 | - | 2, 3 | 178 | 7317608 | ||

Zc3h14
Genomic Map