siRNA Details
| Gene Symbol: Nos1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138922.1 | NP_620277.1 | 10 | 2332 | ||
| GenBank mRNA | AF037071.1 | - | 10 | 2332 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Nos1ap| Gene Symbol: Nos1ap | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138922.1 | NP_620277.1 | 10 | 2332 | ||
| GenBank mRNA | AF037071.1 | - | 10 | 2332 | ||