siRNA Details
Gene Symbol: RGD621098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139040.1 | NP_620609.1 | 3 | 416 | ||
GenBank mRNA | U92010.1 | - | 3 | 416 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: RGD621098 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_139040.1 | NP_620609.1 | 3 | 416 | ||
GenBank mRNA | U92010.1 | - | 3 | 416 |