siRNA Details
| Gene Symbol: Phrf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139093.1 | NP_620793.1 | 13 | 1943 | ||
| XM_006230510.2 | XP_006230572.1 | 14 | 2111 | |||
| XM_006230511.2 | XP_006230573.1 | 14 | 2069 | |||
| XM_006230513.2 | XP_006230575.1 | 14 | 2108 | |||
| XM_006230514.2 | XP_006230576.1 | 14 | 2111 | |||
| XM_006230515.2 | XP_006230577.1 | 14 | 2111 | |||
| XM_006230516.2 | XP_006230578.1 | 14 | 2111 | |||
| XM_006230517.2 | XP_006230579.1 | 14 | 2108 | |||
| GenBank mRNA | U49057.1 | - | 13 | 1943 | ||

Phrf1
Genomic Map