siRNA Details
| Gene Symbol: Scn3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139097.3 | NP_620797.1 | 4 | 691 | ||
| XM_006243102.1 | XP_006243164.1 | 4 | 773 | |||
| XM_006243103.1 | XP_006243165.1 | 4 | 773 | |||
| XM_008766114.1 | XP_008764336.1 | 4 | 773 | |||
| XM_008766115.1 | XP_008764337.1 | 3 | 340 | |||
| GenBank mRNA | AF378093.1 | - | 4 | 390 | ||
| AJ243395.2 | - | 4 | 676 | |||
| BC070899.1 | - | 4 | 691 | 7100476 | ||

Scn3b
Genomic Map