siRNA Details
| Gene Symbol: Dmgdh | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139102.2 | NP_620802.2 | 4 | 447 | ||
| XM_008760629.1 | XP_008758851.1 | 4 | 462 | |||
| XM_008760630.1 | XP_008758852.1 | 4 | 462 | |||
| XM_008760631.1 | XP_008758853.1 | 4 | 462 | |||
| XM_008760632.1 | XP_008758854.1 | 4 | 462 | |||
| GenBank mRNA | BC085697.1 | - | 4 | 447 | 7108858 | |
| X55995.1 | - | 4 | 415 | |||

Dmgdh
Genomic Map