siRNA Details
| Gene Symbol: Egfl7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_139104.1 | NP_620804.1 | 10 | 1489 | ||
| XM_006233668.2 | XP_006233730.1 | 10 | 1591 | |||
| XM_006233669.2 | XP_006233731.1 | 9 | 1145 | |||
| GenBank mRNA | AF223678.1 | - | 10 | 1489 | ||
| BC078725.1 | - | 10 | 1432 | 7127696 | ||

Egfl7
Genomic Map